Learn More
Medchemexpress LLC Coumarin-C2-TCO | 2488174-16-7 | >98.0% | 455.55 | C25H33N3O5 | 10 MG
SDP
Catalog No. 5000287893
Encompass_Preferred
Coumarin-C2-TCO is a coumarin-derived fluorescent dye functionalized with a trans-cyclooctene (TCO) reactive handle that undergoes rapid inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine-bearing molecules for bioorthogonal labeling and conjugation.
  • Fluorescent coumarin core provides a dye scaffold for imaging and detection.
  • TCO reactive handle enables fast, bioorthogonal iEDDA conjugation to tetrazines.
  • Suitable for labeling biomolecules and developing fluorescent probes in chemical biology workflows.
  • Offered in small research quantities for synthesis and assay development.
Catalog No. 50-002-87893 Supplier Medchemexpress LLC Supplier No. HY15630410MG
Coumarin-C2-TCO is a coumarin-derived fluorescent dye functionalized with a trans-cyclooctene (TCO) reactive handle that undergoes rapid inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine-bearing molecules for bioorthogonal labeling and conjugation.
  • Fluorescent coumarin core provides a dye scaffold for imaging and detection.
  • TCO reactive handle enables fast, bioorthogonal iEDDA conjugation to tetrazines.
  • Suitable for labeling biomolecules and developing fluorescent probes in chemical biology workflows.
  • Offered in small research quantities for synthesis and assay development.