Learn More
Medchemexpress LLC Coumarin-d4 | 185056-83-1 | 150.17 | 5 MG
SDP
Catalog No. 5000417111
Encompass_Preferred
Coumarin-d4 is the deuterium labeled version of Coumarin, which is the primary bioactive ingredient found in Radix Glehniae, also known as Beishashen in China. It exhibits various pharmacological activities, including anticancer, anti-inflammation, and antiviral properties. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-17111 Supplier Medchemexpress LLC Supplier No. HYN0709S5MG
Coumarin-d4 is the deuterium labeled version of Coumarin, which is the primary bioactive ingredient found in Radix Glehniae, also known as Beishashen in China. It exhibits various pharmacological activities, including anticancer, anti-inflammation, and antiviral properties. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.