Learn More
Medchemexpress LLC Coumarin-d4 | 185056-83-1 | MFCD01861662 | 150.17 | 1 MG
SDP
Catalog No. 5000417112
Encompass_Preferred
Coumarin-d4 is the deuterium labeled Coumarin. Coumarin is the primary bioactive ingredient in Radix Glehniae, also known as Beishashen in China, which exhibits various pharmacological activities including anticancer, anti-inflammation, and antivirus properties.
  • Deuterium labeled coumarin.
  • Primary bioactive ingredient in Radix Glehniae.
  • Possesses anticancer, anti-inflammation and antivirus activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-17112 Supplier Medchemexpress LLC Supplier No. HYN0709S1MG
Coumarin-d4 is the deuterium labeled Coumarin. Coumarin is the primary bioactive ingredient in Radix Glehniae, also known as Beishashen in China, which exhibits various pharmacological activities including anticancer, anti-inflammation, and antivirus properties.
  • Deuterium labeled coumarin.
  • Primary bioactive ingredient in Radix Glehniae.
  • Possesses anticancer, anti-inflammation and antivirus activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.