Learn More
Cayman Chemical Coumarin hydrazine, Purity: ≥90%, CAS: 113707-87-2, 25 mg
SDP
Catalog No. 502981896
Encompass_Preferred
An aromatic hydrazine-containing fluorophore that reacts with aldehydes or ketones for fluorescent labeling; acts as a sensor for oxidative stress by detection of carbonylated biomolecules in live cells; ex/em = 365/430-550 nm; produces a red shift in emission upon hydrozone formation; displays a large Stokes shift of approximately 195 nm
Catalog No. 50-298-1896 Supplier Cayman Chemical Supplier No. 1911125 MG
An aromatic hydrazine-containing fluorophore that reacts with aldehydes or ketones for fluorescent labeling; acts as a sensor for oxidative stress by detection of carbonylated biomolecules in live cells; ex/em = 365/430-550 nm; produces a red shift in emission upon hydrozone formation; displays a large Stokes shift of approximately 195 nm