Learn More
Medchemexpress LLC Coumarin hydrazine | 113707-87-2 | MFCD09999551 | 97.1% | 190.20 g/mol | C10H10N2O2 | 50MG
SDP
Catalog No. 5000236090
Encompass_Preferred
Coumarin hydrazine is an aromatic hydrazine-containing coumarin fluorophore used as a chemical probe to label protein- and lipid-bound carbonyls in cells. It excites in the 420-450 nm range and emits near 468-490 nm, and is provided as a solid reagent for research applications.
Catalog No. 50-002-36090 Supplier Medchemexpress LLC Supplier No. HY12419750MG
Coumarin hydrazine is an aromatic hydrazine-containing coumarin fluorophore used as a chemical probe to label protein- and lipid-bound carbonyls in cells. It excites in the 420-450 nm range and emits near 468-490 nm, and is provided as a solid reagent for research applications.