Learn More
Medchemexpress LLC Coumarin (standard) | 91-64-5 | 146.14 | 50 MG
SDP
Catalog No. 5000416666
Encompass_Preferred
Coumarin (Standard) is the analytical standard of Coumarin, intended for research and analytical applications. It is a potent and orally active anti-inflammatory agent, and also shows antinociceptive, antibacterial, antifungal, and anticancer activity. This product is for research use only and not sold to patients.
  • The compound is a grade of analytical standard, serving as a reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: phenylpropanoids, coumarins
  • Initial source: plants (Compositae Ambrosia psilostachya DC., Umbelliferae), microorganisms.
Catalog No. 50-004-16666 Supplier Medchemexpress LLC Supplier No. HYN0709R50MG
Coumarin (Standard) is the analytical standard of Coumarin, intended for research and analytical applications. It is a potent and orally active anti-inflammatory agent, and also shows antinociceptive, antibacterial, antifungal, and anticancer activity. This product is for research use only and not sold to patients.
  • The compound is a grade of analytical standard, serving as a reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Appearance: solid
  • Color: white to off-white
  • Structure classification: phenylpropanoids, coumarins
  • Initial source: plants (Compositae Ambrosia psilostachya DC., Umbelliferae), microorganisms.