Learn More
Cayman Chemical CoumarInQuInon ConjugAT 500ug
SDP
Catalog No. 502989805
Encompass_Preferred
A fluorescent substrate for NADH:ubiquinone oxidoreductases; comprised of a coumarin fluorophore conjugated to a ubiquinone analog that can be reduced by NADH:ubiquinone oxidoreductases; has been used to measure the kinetic parameters of AIFM2/FSP1
Catalog No. 50-298-9805 Supplier Cayman Chemical Supplier No. 29554500 UG
A fluorescent substrate for NADH:ubiquinone oxidoreductases; comprised of a coumarin fluorophore conjugated to a ubiquinone analog that can be reduced by NADH:ubiquinone oxidoreductases; has been used to measure the kinetic parameters of AIFM2/FSP1