Learn More
Enzo Life Sciences Coumestrol (5 mg). CAS: 479-13-0
SDP
Catalog No. 502011742
Encompass_Preferred
Phytoestrogen. Acts as an estrogen agonist. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml) and warm ethanol (5 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1742 Supplier Enzo Life Sciences Supplier No. BMLS1800005
Phytoestrogen. Acts as an estrogen agonist. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml) and warm ethanol (5 mg/ml). Long Term Storage: Ambient.