Learn More
Medchemexpress LLC HY-N2335 5mg Medchemexpress, Coumestrol CAS:479-13-0 Purity:>98%
SDP
Catalog No. 501933914
Encompass_Preferred
Medchemexpress, HY-N2335 5mg Coumestrol CAS:479-13-0 Coumestrol, a phytoestrogen present in soybean products, exhibits activities against cancers, neurological disorders, and autoimmune diseases. It suppresses proliferation of ES2 cells with an IC 50 of 50 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3914 Supplier Medchemexpress LLC Supplier No. HYN23355MG
Medchemexpress, HY-N2335 5mg Coumestrol CAS:479-13-0 Coumestrol, a phytoestrogen present in soybean products, exhibits activities against cancers, neurological disorders, and autoimmune diseases. It suppresses proliferation of ES2 cells with an IC 50 of 50 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.