Learn More
World Precision Instrument Councilman Rib Shears Box Lock, 11 in
SDP
Catalog No. 502538378
Encompass
The Councilman Rib Shears are used to cut (or break) bone. The long handles give you more leverage, making the action easier to accomplish. The blades are sharp, and the spring action keeps them open till you are ready to use them. These shears are crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8378 Supplier World Precision Instrument Supplier No. WP0049
The Councilman Rib Shears are used to cut (or break) bone. The long handles give you more leverage, making the action easier to accomplish. The blades are sharp, and the spring action keeps them open till you are ready to use them. These shears are crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.