Learn More
Electron Microscopy Sciences Count Up/Count Down Timer Certified
SDP
Catalog No. 50949423
Encompass_Preferred
Count up or down for 100 hours in intervals of 1 second. In the count down mode its unique feature tells you how much time has elapsed after the alarm sounds. Its memory allows for the repeat of a cycle just completed. Alarm clock mode. It comes complete with a built in stand, pocket clip, button cell battery, and magnet. With N.I.S.T. traceable certificate of compliance. NIST traceable certificate
Catalog No. 50-949-423 Supplier Electron Microscopy Sciences Supplier No. 7260302
Count up or down for 100 hours in intervals of 1 second. In the count down mode its unique feature tells you how much time has elapsed after the alarm sounds. Its memory allows for the repeat of a cycle just completed. Alarm clock mode. It comes complete with a built in stand, pocket clip, button cell battery, and magnet. With N.I.S.T. traceable certificate of compliance. NIST traceable certificate