Learn More
Med Vet International Count Right Pill Counter, J1245N, Each
Supplier:  Med Vet International JOR1245N
Encompass_Preferred
Counting pills is an ardous task for every clinic. It is a time consuming and expensive job that no one wants. Other electronic pill counters are fast, accurate, and convenient, but cost thousands of dollars. The JorVet Count-Right Pill Counter is a simple design at a moderate price. The counter uses precise weighing based on the initial weighing of an empty pill vial followed by a 5 pill weight count. Then the desired pill count balance is added. - Counts all size pills and capsules even extra-large ‘horse' tablets. - AC plug in that converts to 6V DC. - Simple on/off switch or shuts off automatically after 30 seconds of non-use. - Easy and simple to learn how to use. Dimensions: 41?2"W x 41?2"L x 21?4"H Weight: 11oz (330gm) Warranty: Two years on manufacturer
Catalog No. 50-004-95792