Learn More
Med Vet International Countersinks, Bone Screw, J0372Y, Large, Each
Catalog No. 5000581841
Encompass_Preferred
This instrument contours the top of the screw hole to fit the screwhead. It is used for lag screw procedures and for the reattachment of small bone fragments.
Catalog No. 50-005-81841 Supplier Med Vet International Supplier No. JOR372Y
This instrument contours the top of the screw hole to fit the screwhead. It is used for lag screw procedures and for the reattachment of small bone fragments.