Learn More
Poltex Tilted Countertop Glove Box Holder
Description
- Holder is designed with a backwards tilt for easy dispensing and stability on any flat surface
- Constructed from durable PETG which features higher chemical resistance than acrylic
- Rubber bumpons on the base prevent scratching and slipping
- Top loading design is compatible with virtually any size glove box
- Gloves are sold separately
Specifications
Specifications
| Product Type | Glove Box Holder |
| Color | Clear |
| Dimensions (D x W x H) Interior | 3.5 x 23 x 9 in. |
| Holds | Disposable Glove Boxes |
| Material | PETG |
| Mounting | Freestanding |
| No. of Compartments | 1 |
| Product Style | Top Loading |
| Capacity | 4 Boxes |