Learn More
Grainger A&D Weighing Industrial Bench Scale, 15 lb Capacity, Stainless Steel Weighing Pan, 1 ea
Catalog No. 502779770
Encompass_Preferred
The HC-6Ki industrial counting scale is designed for precise parts counting, weighing, and check-weighing applications. Featuring a robust plastic housing and a stainless steel weighing pan, this unit includes a large, detachable 3-window LCD display that provides clear visibility of piece, weight, and unit weight data. It offers an internal resolution of up to 1/750,000 and supports advanced counting accuracy through the Automatic Counting Accuracy Improvement (ACAI) function.
  • Up to 99 unit weight ID memory function for efficient data retrieval.
  • Audible weighing assist beep function for reaching target quantities.
  • Integrated level bubble and adjustable leveling feet for proper setup.
  • Comparator function with HI, OK, and LO indicators for check-weighing.
  • Auto power off function to conserve energy.
  • Portable design with optional battery operation up to 80 hr.
Catalog No. 50-277-9770 Supplier Grainger Supplier No. 19ND05
The HC-6Ki industrial counting scale is designed for precise parts counting, weighing, and check-weighing applications. Featuring a robust plastic housing and a stainless steel weighing pan, this unit includes a large, detachable 3-window LCD display that provides clear visibility of piece, weight, and unit weight data. It offers an internal resolution of up to 1/750,000 and supports advanced counting accuracy through the Automatic Counting Accuracy Improvement (ACAI) function.
  • Up to 99 unit weight ID memory function for efficient data retrieval.
  • Audible weighing assist beep function for reaching target quantities.
  • Integrated level bubble and adjustable leveling feet for proper setup.
  • Comparator function with HI, OK, and LO indicators for check-weighing.
  • Auto power off function to conserve energy.
  • Portable design with optional battery operation up to 80 hr.