Learn More
Grainger A&D Engineering HD Series Counting Scale, 60 lb Capacity, 1 ea
Catalog No. 502779786
Encompass_Preferred
This high-capacity counting scale is designed for precise weighing and inventory tasks, featuring an internal resolution of 1/120,000 for accurate piece counting. It utilizes advanced technology to minimize errors in unit weight variation and includes a detachable display for flexible operation in various environments.
  • Capacity of 27.2 kg (60 lb.) with a display resolution of 0.005 kg (0.01 lb.).
  • Includes Automatic Count Accuracy Improvement (ACAI) to correct unit weight errors.
  • Equipped with a comparator function with hi/ok/lo display for packing applications.
  • Features a large 3-window LCD display with 17 mm (0.67 in.) character height.
  • Offers versatile power options including AC adapter, 6 C-cell batteries, or optional rechargeable NiCd battery pack.
  • Memory plus function for accumulating count data.
  • Wide operating temperature range of -10°C to 40°C.
Catalog No. 50-277-9786 Supplier Grainger Supplier No. 19ND35
This high-capacity counting scale is designed for precise weighing and inventory tasks, featuring an internal resolution of 1/120,000 for accurate piece counting. It utilizes advanced technology to minimize errors in unit weight variation and includes a detachable display for flexible operation in various environments.
  • Capacity of 27.2 kg (60 lb.) with a display resolution of 0.005 kg (0.01 lb.).
  • Includes Automatic Count Accuracy Improvement (ACAI) to correct unit weight errors.
  • Equipped with a comparator function with hi/ok/lo display for packing applications.
  • Features a large 3-window LCD display with 17 mm (0.67 in.) character height.
  • Offers versatile power options including AC adapter, 6 C-cell batteries, or optional rechargeable NiCd battery pack.
  • Memory plus function for accumulating count data.
  • Wide operating temperature range of -10°C to 40°C.