Learn More
YMC America, Inc COUPLER 1/16IN OD .02IN ID 5CM
SDP
Catalog No. 50249555
Encompass
XRCP1605: Column Coupler, 1/16 in. OD, 0.02 in. ID, 5cm Length
Catalog No. 50-249-555 Supplier YMC America, Inc Supplier No. XRCP1605
XRCP1605: Column Coupler, 1/16 in. OD, 0.02 in. ID, 5cm Length