Learn More
Mac-Mod PEEK Column Coupler for All-Guard Cartridge Holder (standard column port)
SDP
Catalog No. 5090002072
Encompass
The PEEK one-piece fingertight column coupler is ideal for connecting two analytical columns or analytical columns to guard columns or cartridges. Ultra low dead volume. Inert and biocompatible. Withstands pressure up to 6000 psi. Female-female connector.
Catalog No. 50-900-02072 Supplier Mac-Mod Supplier No. HI081
The PEEK one-piece fingertight column coupler is ideal for connecting two analytical columns or analytical columns to guard columns or cartridges. Ultra low dead volume. Inert and biocompatible. Withstands pressure up to 6000 psi. Female-female connector.