Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 10/PK
Catalog No. 503131352
Encompass_Preferred
CPC HFC Series High-Volume Quick-Disconnect Coupling Bodies Diameter Inner 0 5 in 9 5 mm Overall Length 2 8 in 71 2 mm Thread Size 0 5 in Barb valved Gray For Use With 12 7mm Includes Shut-off valves EPDM Seal Hose Barb Body Height 1 4 in 36 mm Maximum Pressure 60 psi
Catalog No. 50-313-1352 Supplier COLDER PRODUCTS COMPANY Supplier No. HFCD17812
CPC HFC Series High-Volume Quick-Disconnect Coupling Bodies Diameter Inner 0 5 in 9 5 mm Overall Length 2 8 in 71 2 mm Thread Size 0 5 in Barb valved Gray For Use With 12 7mm Includes Shut-off valves EPDM Seal Hose Barb Body Height 1 4 in 36 mm Maximum Pressure 60 psi