Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 25/PK
Catalog No. 503131282
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 9 5mm Polypropylene Overall Length 49 7 mm 1 96 in Almond Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 1 02 in 26 mm Maximum Pressure 120 psi
Catalog No. 50-313-1282 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD1700612
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 9 5mm Polypropylene Overall Length 49 7 mm 1 96 in Almond Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 1 02 in 26 mm Maximum Pressure 120 psi