Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 50/PK
Catalog No. 503131338
Encompass_Preferred
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 1 6mm Polypropylene Hose Barb Non-valved Coupling Insert Autoclavable Not Autoclavable White Overall Length 0 75 in 19 mm Height 0 48 in 12 1 mm Maximum Pressure 100 psi
Catalog No. 50-313-1338 Supplier COLDER PRODUCTS COMPANY Supplier No. SMM0112
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 1 6mm Polypropylene Hose Barb Non-valved Coupling Insert Autoclavable Not Autoclavable White Overall Length 0 75 in 19 mm Height 0 48 in 12 1 mm Maximum Pressure 100 psi