Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 50/PK
Catalog No. 503131397
Encompass_Preferred
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 3 2mm Acetal Hose Barb Non-Valved In-line Coupling Body Autoclavable Autoclavable White Overall Length 0 86 in 21 8 mm Height 0 51 in 13 mm Maximum Pressure 100 psi
Catalog No. 50-313-1397 Supplier COLDER PRODUCTS COMPANY Supplier No. SMPT02
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 3 2mm Acetal Hose Barb Non-Valved In-line Coupling Body Autoclavable Autoclavable White Overall Length 0 86 in 21 8 mm Height 0 51 in 13 mm Maximum Pressure 100 psi