Learn More
COLDER PRODUCTS COMPANY COUPLER BODY 50/PK
Supplier:  COLDER PRODUCTS COMPANY SMF01
Encompass_Preferred
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 1 6mm Acetal Hose Barb Non-Valved In-line Coupling Body Autoclavable Autoclavable White Overall Length 0 75 to 0 9 in 19 to 22 mm Height 0 48 in 12 1 mm
Catalog No. 50-313-1429