Learn More
COLDER PRODUCTS COMPANY COUPLER BODY EA
Catalog No. 503131388
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 6 4mm EPDM Seal Overall Length 49 7 mm 1 96 in Almond Diameter Inner 0 25 in Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 1 02 in 26 mm Maximum Pressure 120 psi
Catalog No. 50-313-1388 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD1700412
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 6 4mm EPDM Seal Overall Length 49 7 mm 1 96 in Almond Diameter Inner 0 25 in Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 1 02 in 26 mm Maximum Pressure 120 psi