Learn More
IDEX Health & Science Upchurch COUPLER FINGERTIGHT II
Catalog No. NC1920530
Encompass_Preferred
Idex Universal Column Couplers, For Use With: F-200 Derlin Fingertights w/0.007 in. I.D., Connect Column w/Internal 10-32 Threads to Precolumn or Guard Column w/10-32 Threads, Swept Volume: 1.3 uL
Catalog No. NC1920530 Supplier IDEX Health & Science Upchurch Supplier No. U284
Idex Universal Column Couplers, For Use With: F-200 Derlin Fingertights w/0.007 in. I.D., Connect Column w/Internal 10-32 Threads to Precolumn or Guard Column w/10-32 Threads, Swept Volume: 1.3 uL