Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 10/PK
Supplier:  COLDER PRODUCTS COMPANY HFCD10612
Encompass_Preferred
CPC HFC Series High-Volume Quick-Disconnect Coupling Bodies Diameter Inner 0 375 in 9 5 mm Overall Length 2 7 in 68 5 mm Thread Size 0 375 in NPT Gray For Use With 9 5mm Includes Shut-off valves EPDM Seal MNPT Pipe-thread Body Height 1 4 in 36 mm Maximum Pressure 60 psi
Catalog No. 50-313-1301