Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 10/PK
Catalog No. 503131302
Encompass_Preferred
CPC HFC Series High-Volume Quick-Disconnect Coupling Inserts With Shut-Off Valve Diameter Inner 0 5 in 9 5 mm MNPT Pipe-thread Insert Height 1 16 in 29 4 mm Overall Length 2 14 in 52 mm Gray For Use With 12 7mm Includes Shut-off valves Polypropylene Maximum Pressure 60 psi
Catalog No. 50-313-1302 Supplier COLDER PRODUCTS COMPANY Supplier No. HFCD24812
CPC HFC Series High-Volume Quick-Disconnect Coupling Inserts With Shut-Off Valve Diameter Inner 0 5 in 9 5 mm MNPT Pipe-thread Insert Height 1 16 in 29 4 mm Overall Length 2 14 in 52 mm Gray For Use With 12 7mm Includes Shut-off valves Polypropylene Maximum Pressure 60 psi