Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 10/PK
Catalog No. 503131315
Encompass_Preferred
CPC HFC Series High-Volume Quick-Disconnect Coupling Inserts With Straight-Through Flow For Use With 12 7mm Straight Hose-barb Insert Height 1 in 25 4 mm Overall Length 1 9 in 50 mm Gray Diameter Inner 0 5 in 9 5 mm EPDM Seal Maximum Pressure 60 psi
Catalog No. 50-313-1315 Supplier COLDER PRODUCTS COMPANY Supplier No. HFC22812
CPC HFC Series High-Volume Quick-Disconnect Coupling Inserts With Straight-Through Flow For Use With 12 7mm Straight Hose-barb Insert Height 1 in 25 4 mm Overall Length 1 9 in 50 mm Gray Diameter Inner 0 5 in 9 5 mm EPDM Seal Maximum Pressure 60 psi