Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 10/PK
Supplier: COLDER PRODUCTS COMPANY HFC22612
CPC HFC Series High-Volume Quick-Disconnect Coupling Inserts With Straight-Through Flow For Use With 9 5mm Fits into Intramedic PE 50 tubing Straight Hose-barb Insert Height 1 in 25 4 mm Overall Length 1 9 in 50 mm Gray Diameter Inner 0 375 in 9 5 mm EPDM Seal