Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Catalog No. 503131311
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PLC Series Buna-N Seal For Use With 6 4mm Max Operating Temperature 82 deg C 180 deg F White Diameter Inner 0 25 mm Includes Shut-off valves
Catalog No. 50-313-1311 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD22004
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PLC Series Buna-N Seal For Use With 6 4mm Max Operating Temperature 82 deg C 180 deg F White Diameter Inner 0 25 mm Includes Shut-off valves