Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Catalog No. 503131283
Encompass_Preferred
CPC Ferruleless Polytube Inserts with Shut-Off Valve White Buna-N Seal Height 0 72 in 18 2 mm Overall Length 1 3 to 1 8 in 33 to 45 mm For Use With 6 4mm Includes Shut-off valves PTF Valved In-Line Coupling Insert Maximum Pressure 120 psi
Catalog No. 50-313-1283 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD20006
CPC Ferruleless Polytube Inserts with Shut-Off Valve White Buna-N Seal Height 0 72 in 18 2 mm Overall Length 1 3 to 1 8 in 33 to 45 mm For Use With 6 4mm Includes Shut-off valves PTF Valved In-Line Coupling Insert Maximum Pressure 120 psi