Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Catalog No. 503131337
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 3 2mm For use in laboratory equipment medical and surgical instrumentation water transfer air systems and other applications Buna-N Seal Max Operating Temperature 82 deg C 180 deg F
Catalog No. 50-313-1337 Supplier COLDER PRODUCTS COMPANY Supplier No. PMC2202
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 3 2mm For use in laboratory equipment medical and surgical instrumentation water transfer air systems and other applications Buna-N Seal Max Operating Temperature 82 deg C 180 deg F