Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD2202
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PMC Series Acetal For Use With 3 2mm Max Operating Temperature 82 deg C 180 deg F White Diameter Inner 0 125 mm Includes Shut-off valves Overall Length 1 to 1 67 in 25 to 42 4 mm
Catalog No. 50-313-1276