Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier: COLDER PRODUCTS COMPANY PLC2200612
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 9 5mm EPDM Seal Overall Length 33 to 45 mm 1 3 to 1 8 in Almond Hose Barb Valved In-line Coupling Insert Height 0 63 in 16 mm Maximum Pressure 120 psi