Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD220212
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PMC Series EPDM Seal For Use With 3 2mm Max Operating Temperature 71 deg C 160 deg F Almond Diameter Inner 0 125 mm Includes Shut-off valves
Catalog No. 50-313-1386