Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Catalog No. 503131408
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PLC Series Acetal For Use With 9 5mm Max Operating Temperature 82 deg C 180 deg F White Diameter Inner 0 37 mm Includes Shut-off valves
Catalog No. 50-313-1408 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD22006
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PLC Series Acetal For Use With 9 5mm Max Operating Temperature 82 deg C 180 deg F White Diameter Inner 0 37 mm Includes Shut-off valves