Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PLC2200412
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 6 4mm EPDM Seal Max Operating Temperature 71 deg C 160 deg F Min Operating Temperature 0 deg C 32 deg F Almond Diameter Inner 0 25 mm Overall Length 1 35 to 2 in 34 2 to 50 8 mm
Catalog No. 50-313-1440