Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD200412
Encompass_Preferred
CPC Ferruleless Polytube Inserts with Shut-Off Valve Almond EPDM Seal Height 0 58 in 14 7 mm Overall Length 1 15 to 1 58 in 29 to 40 mm Diameter Inner 0 125 in For Use With 4 3mm Includes Shut-off valves PTF Valved In-Line Coupling Insert Maximum Pressure 120 psi
Catalog No. 50-313-1444