Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 50/PK
Supplier:  COLDER PRODUCTS COMPANY SMF02
Encompass_Preferred
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 3 2mm Acetal Hose Barb Non-Valved In-line Coupling Body Autoclavable Autoclavable White Overall Length 0 9 in 22 mm Height 0 48 in 12 1 mm Max Operating Temperature 180 deg F 82 deg C
Catalog No. 50-313-1360