Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 50/PK
Catalog No. 503131277
Encompass_Preferred
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 3 2mm Acetal Hose Barb Non-valved Coupling Insert Autoclavable Autoclavable White Overall Length 0 9 in 22 mm Height 0 48 in 12 1 mm Maximum Pressure 100 psi
Catalog No. 50-313-1277 Supplier COLDER PRODUCTS COMPANY Supplier No. SMM02
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 3 2mm Acetal Hose Barb Non-valved Coupling Insert Autoclavable Autoclavable White Overall Length 0 9 in 22 mm Height 0 48 in 12 1 mm Maximum Pressure 100 psi