Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 50/PK
Supplier:  COLDER PRODUCTS COMPANY SMM0212
Encompass_Preferred
CPC SMC Series Subminiature Quick-Disconnect Coupling Bodies and Inserts For Use With 3 2mm Buna-N Seal Hose Barb Non-valved Coupling Insert Autoclavable Not Autoclavable White Overall Length 0 9 in 22 mm Height 0 48 in 12 1 mm Maximum Pressure 100 psi
Catalog No. 50-313-1359