Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT EA
Catalog No. 503131278
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 9 5mm Acetal Max Operating Temperature 82 deg C 180 deg F Min Operating Temperature -40 deg C -40 deg F White Diameter Inner 0 37 mm Overall Length 1 36 to 1 81 in 34 5 to 46 mm
Catalog No. 50-313-1278 Supplier COLDER PRODUCTS COMPANY Supplier No. PLC22006
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 9 5mm Acetal Max Operating Temperature 82 deg C 180 deg F Min Operating Temperature -40 deg C -40 deg F White Diameter Inner 0 37 mm Overall Length 1 36 to 1 81 in 34 5 to 46 mm