Learn More
Instech Stainless steel catheter / tubing coupler, 17ga x 15mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081202
Encompass
The Instech SC17/15 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 17ga and 15mm in length, it provides a secure, low‑dead‑volume connection for infusion or sampling setups. The coupler’s rounded edges help prevent tubing damage, and it is supplied non‑sterile in packages of 100 for use in assembling or repairing infusion lines.
Features
  • 17ga stainless steel straight catheter coupler
  • 15mm length for secure tubing engagement
  • Rounded edges to minimize catheter or tubing damage
  • Non‑sterile; supplied in packs of 100
  • Connects catheters to external infusion tubing
Catalog No. 50-812-02 Supplier Instech Supplier No. SC17/15
The Instech SC17/15 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 17ga and 15mm in length, it provides a secure, low‑dead‑volume connection for infusion or sampling setups. The coupler’s rounded edges help prevent tubing damage, and it is supplied non‑sterile in packages of 100 for use in assembling or repairing infusion lines.
Features
  • 17ga stainless steel straight catheter coupler
  • 15mm length for secure tubing engagement
  • Rounded edges to minimize catheter or tubing damage
  • Non‑sterile; supplied in packs of 100
  • Connects catheters to external infusion tubing