Learn More
Instech Stainless steel catheter / tubing coupler, 20ga x 15mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081203
Encompass
The Instech SC20/15 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 20ga and 15mm in length, it provides a secure, low‑dead‑volume junction for reliable fluid transfer in infusion or sampling setups. The rounded edges reduce the risk of tubing damage, and the couplers are supplied non‑sterile in packages of 100 for laboratory use.
Features
  • 20ga stainless steel straight catheter coupler
  • 15mm length for secure catheter‑to‑tubing connection
  • Rounded edges to protect tubing during use
  • Non‑sterile; packaged in sets of 100
  • Used to connect catheters to external infusion lines
Catalog No. 50-812-03 Supplier Instech Supplier No. SC20/15
The Instech SC20/15 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 20ga and 15mm in length, it provides a secure, low‑dead‑volume junction for reliable fluid transfer in infusion or sampling setups. The rounded edges reduce the risk of tubing damage, and the couplers are supplied non‑sterile in packages of 100 for laboratory use.
Features
  • 20ga stainless steel straight catheter coupler
  • 15mm length for secure catheter‑to‑tubing connection
  • Rounded edges to protect tubing during use
  • Non‑sterile; packaged in sets of 100
  • Used to connect catheters to external infusion lines