Learn More
Instech Stainless steel catheter / tubing coupler, 22ga x 15mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081204
Encompass
The Instech SC22/15 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 22ga and 15mm in length, it provides a secure, low‑dead‑volume junction for reliable fluid transfer in rodent infusion and sampling studies. Its rounded edges help prevent tubing damage, and it is supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 22ga stainless steel straight catheter coupler
  • 15mm length for secure catheter‑to‑tubing connection
  • Rounded edges minimize risk of tubing damage
  • Non‑sterile; supplied in packs of 100
  • Connects implanted catheters to external infusion lines
Catalog No. 50-812-04 Supplier Instech Supplier No. SC22/15
The Instech SC22/15 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 22ga and 15mm in length, it provides a secure, low‑dead‑volume junction for reliable fluid transfer in rodent infusion and sampling studies. Its rounded edges help prevent tubing damage, and it is supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 22ga stainless steel straight catheter coupler
  • 15mm length for secure catheter‑to‑tubing connection
  • Rounded edges minimize risk of tubing damage
  • Non‑sterile; supplied in packs of 100
  • Connects implanted catheters to external infusion lines