Learn More
Instech Stainless steel catheter / tubing coupler, 22ga x 8mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081206
Encompass
The Instech SC22/8 is a stainless steel catheter coupler used to join implanted catheters to external infusion tubing. Sized at 22ga and 8mm in length, it provides a compact, low‑dead‑volume connection suitable for small‑animal infusion setups. The rounded edges help prevent tubing damage, and it is supplied non‑sterile in packages of 100 for laboratory use.
Features
  • 22ga stainless steel straight catheter coupler
  • 8mm length for compact catheter‑to‑tubing connections
  • Rounded edges to minimize tubing wear
  • Non‑sterile; packaged in sets of 100
  • Used to connect implanted catheters to external infusion lines
Catalog No. 50-812-06 Supplier Instech Supplier No. SC22/8
The Instech SC22/8 is a stainless steel catheter coupler used to join implanted catheters to external infusion tubing. Sized at 22ga and 8mm in length, it provides a compact, low‑dead‑volume connection suitable for small‑animal infusion setups. The rounded edges help prevent tubing damage, and it is supplied non‑sterile in packages of 100 for laboratory use.
Features
  • 22ga stainless steel straight catheter coupler
  • 8mm length for compact catheter‑to‑tubing connections
  • Rounded edges to minimize tubing wear
  • Non‑sterile; packaged in sets of 100
  • Used to connect implanted catheters to external infusion lines