Learn More
Instech Stainless steel catheter / tubing coupler, 23ga x 8mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081207
Encompass
The Instech SC23/8 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 23ga and 8mm in length, it provides a compact, secure, low‑dead‑volume junction suitable for small‑animal infusion and sampling workflows. The rounded edges protect tubing from damage, and the couplers are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 23ga stainless steel straight catheter coupler
  • 8mm length for compact catheter‑to‑tubing connections
  • Rounded edges to minimize tubing wear
  • Non‑sterile; packaged in sets of 100
  • Connects implanted catheters to external infusion lines
Catalog No. 50-812-07 Supplier Instech Supplier No. SC23/8
The Instech SC23/8 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 23ga and 8mm in length, it provides a compact, secure, low‑dead‑volume junction suitable for small‑animal infusion and sampling workflows. The rounded edges protect tubing from damage, and the couplers are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 23ga stainless steel straight catheter coupler
  • 8mm length for compact catheter‑to‑tubing connections
  • Rounded edges to minimize tubing wear
  • Non‑sterile; packaged in sets of 100
  • Connects implanted catheters to external infusion lines