Learn More
Instech Stainless steel catheter / tubing coupler, 25ga x 10mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081208
Encompass
The Instech SC25/10 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 25ga and 10mm in length, it provides a secure, low‑dead‑volume junction ideal for small‑animal infusion and sampling procedures. Its rounded edges help prevent tubing damage, and it is supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 25ga stainless steel straight catheter coupler
  • 10mm length for compact catheter‑to‑tubing connections
  • Rounded edges to minimize tubing wear
  • Non‑sterile; packaged in sets of 100
  • Connects implanted catheters to external infusion lines
Catalog No. 50-812-08 Supplier Instech Supplier No. SC25/10
The Instech SC25/10 is a stainless steel catheter coupler used to connect implanted catheters to external infusion tubing. Sized at 25ga and 10mm in length, it provides a secure, low‑dead‑volume junction ideal for small‑animal infusion and sampling procedures. Its rounded edges help prevent tubing damage, and it is supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 25ga stainless steel straight catheter coupler
  • 10mm length for compact catheter‑to‑tubing connections
  • Rounded edges to minimize tubing wear
  • Non‑sterile; packaged in sets of 100
  • Connects implanted catheters to external infusion lines