Learn More
Instech Stainless steel catheter / tubing coupler, 27ga x 8mm. Nonsterile. Package of 100.
SDP
Catalog No. 5081209
Encompass
The Instech SC27/8 is a stainless steel catheter coupler designed to connect implanted catheters to external infusion tubing. Sized at 27ga and 8mm in length, it provides an ultra‑fine, low‑dead‑volume connection ideal for small‑animal or micro‑catheter applications. The rounded edges help protect delicate tubing, and the couplers are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 27ga stainless steel straight catheter coupler
  • 8mm length for compact micro‑tubing connections
  • Rounded edges to minimize tubing wear or tearing
  • Non‑sterile; packaged in sets of 100
  • Connects implanted micro‑catheters to external infusion lines
Catalog No. 50-812-09 Supplier Instech Supplier No. SC27/8
The Instech SC27/8 is a stainless steel catheter coupler designed to connect implanted catheters to external infusion tubing. Sized at 27ga and 8mm in length, it provides an ultra‑fine, low‑dead‑volume connection ideal for small‑animal or micro‑catheter applications. The rounded edges help protect delicate tubing, and the couplers are supplied non‑sterile in packs of 100 for laboratory use.
Features
  • 27ga stainless steel straight catheter coupler
  • 8mm length for compact micro‑tubing connections
  • Rounded edges to minimize tubing wear or tearing
  • Non‑sterile; packaged in sets of 100
  • Connects implanted micro‑catheters to external infusion lines