Learn More
Grainger Coupling 3/4 in CTS PEX White
Catalog No. 502768612
Encompass_Preferred
The 3/4" PEX Coupling by Apollo is made of durable PEX plastic and has a maximum pressure rating of 160 PSI at 73°F. It is designed for use with PEX tubing in potable water applications and connects two sections of 3/4" PEX tubing together. The coupling has a white finish and features stainless steel ring support for enhanced durability. The dimensions are 1.3" length x 1.5" width. This coupling provides a simple push-fit connection and is an ideal solution for creating secure PEX joints.
Catalog No. 50-276-8612 Supplier Grainger Supplier No. 16T738
The 3/4" PEX Coupling by Apollo is made of durable PEX plastic and has a maximum pressure rating of 160 PSI at 73°F. It is designed for use with PEX tubing in potable water applications and connects two sections of 3/4" PEX tubing together. The coupling has a white finish and features stainless steel ring support for enhanced durability. The dimensions are 1.3" length x 1.5" width. This coupling provides a simple push-fit connection and is an ideal solution for creating secure PEX joints.